Learn More
Medchemexpress LLC Metalaxyl | 57837-19-1 | 99.55% | 100 MG
SDP
Catalog No. 5000409396
Encompass_Preferred
Metalaxyl is a fungicide agent that inhibits protein synthesis in fungi. It is effective against downy mildews and soil-borne diseases caused by *Phytium ssp.* and *Phytophthora ssp.*. In vitro, Metalaxyl inhibits the growth of *P. infestans* fungal isolates from Serbian potato fields (EC50s = 0.3-3.9 μg/mL). In vivo, it causes dose-dependent bradycardia in albino rats, with higher doses (250 and 300 mg/kg body weight) leading to sustained bradycardia and cardiac arrest, though acetylcholinesterase activities were not affected.
  • Inhibits protein synthesis in fungi
  • Effective against downy mildews
  • Effective against soil-borne diseases caused by *Phytium ssp.* and *Phytophthora ssp.*
  • Inhibits growth of *P. infestans* fungal isolates in vitro
Catalog No. 50-004-09396 Supplier Medchemexpress LLC Supplier No. HYB0843100MG
Metalaxyl is a fungicide agent that inhibits protein synthesis in fungi. It is effective against downy mildews and soil-borne diseases caused by *Phytium ssp.* and *Phytophthora ssp.*. In vitro, Metalaxyl inhibits the growth of *P. infestans* fungal isolates from Serbian potato fields (EC50s = 0.3-3.9 μg/mL). In vivo, it causes dose-dependent bradycardia in albino rats, with higher doses (250 and 300 mg/kg body weight) leading to sustained bradycardia and cardiac arrest, though acetylcholinesterase activities were not affected.
  • Inhibits protein synthesis in fungi
  • Effective against downy mildews
  • Effective against soil-borne diseases caused by *Phytium ssp.* and *Phytophthora ssp.*
  • Inhibits growth of *P. infestans* fungal isolates in vitro