Learn More
Medchemexpress LLC Metalaxyl | 57837-19-1 | 99.55% | 50 MG
SDP
Catalog No. 5000408732
Encompass_Preferred
Metalaxyl is a fungicide agent that inhibits protein synthesis in fungi. It is effective against downy mildews and soil-borne diseases caused by Phytium ssp. and Phytophthora ssp..
  • Inhibits protein synthesis in fungi.
  • Effective against downy mildews.
  • Effective against soil-borne diseases.
Catalog No. 50-004-08732 Supplier Medchemexpress LLC Supplier No. HYB084350MG
Metalaxyl is a fungicide agent that inhibits protein synthesis in fungi. It is effective against downy mildews and soil-borne diseases caused by Phytium ssp. and Phytophthora ssp..
  • Inhibits protein synthesis in fungi.
  • Effective against downy mildews.
  • Effective against soil-borne diseases.