Learn More
Medchemexpress LLC Metalaxyl-M (R)-metalaxyl (standard) | 70630-17-0 | MFCD03701742 | 97.4% | 279.33 | C15H21NO4 | 10 MG
SDP
Catalog No. 5000297379
Encompass_Preferred
Metalaxyl-M is the analytical standard of (R)-metalaxyl used for research and analytical applications. It is the active (R)-enantiomer of metalaxyl, a fungicide that selectively inhibits fungal RNA polymerase, and is used as a reference standard in chromatography, residue analysis, and ecotoxicology studies.
  • Analytical standard for (R)-metalaxyl
  • Active (R)-enantiomer with fungicidal activity
  • Reference standard for chromatography and residue analysis
  • Suitable for ecotoxicology and hepatocyte metabolism studies
  • Available in small milligram pack sizes for analytical work
Catalog No. 50-002-97379 Supplier Medchemexpress LLC Supplier No. HYB0843AR10MG
Metalaxyl-M is the analytical standard of (R)-metalaxyl used for research and analytical applications. It is the active (R)-enantiomer of metalaxyl, a fungicide that selectively inhibits fungal RNA polymerase, and is used as a reference standard in chromatography, residue analysis, and ecotoxicology studies.
  • Analytical standard for (R)-metalaxyl
  • Active (R)-enantiomer with fungicidal activity
  • Reference standard for chromatography and residue analysis
  • Suitable for ecotoxicology and hepatocyte metabolism studies
  • Available in small milligram pack sizes for analytical work