Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metalaxyl-(methoxyacetyl OMe-d3), Analytical Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001602 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1602 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1602 Supplier MilliporeSigma Supelco Supplier No. 089635MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Line PESTANAL
Linear Formula C15D3H18NO4
Synonym N-(2,6-Dimethylphenyl)-N-(methoxy-d3-acetyl)-DL-alanine methyl ester; Metalaxyl-d4
Quantity 5 mg
Formula Weight 282.35
Percent Purity ≥98.0% (HPLC)
Grade PESTANAL™ Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.