Learn More
Medchemexpress LLC Metaldehyde-d16 | 1219805-73-8 | 500 UG
SDP
Catalog No. 5000412255
Encompass_Preferred
Metaldehyde-d16 is a deuterium labeled Metaldehyde. It can be used as a tracer and as an internal standard for quantitative analysis.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-12255 Supplier Medchemexpress LLC Supplier No. HYB1870S500UG
Metaldehyde-d16 is a deuterium labeled Metaldehyde. It can be used as a tracer and as an internal standard for quantitative analysis.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS