Learn More
Grainger TapeCase Metalized Film Tape, Silver, Polyester, 1 in. x 72 yd.
Catalog No. 502754967
Encompass_Preferred
This metalized polyester film tape provides a reflective surface, offering dimensional stability for various decorative, shielding, and labeling applications. Designed for versatile use, it features a durable construction suitable for indoor and outdoor environments.
  • Features an aggressive, long-lasting acrylic adhesive
  • Offers excellent chemical and thermal stability
  • Includes a brilliant, vibrant, mirror-like finish
  • Compatible with a wide range of surfaces including metal, polycarbonate, and powder-coated paints
  • Withstands temperatures from -60°F to +300°F
  • Provides a tensile strength of 24 lbs./in.
Catalog No. 50-275-4967 Supplier Grainger Supplier No. 15D415
This metalized polyester film tape provides a reflective surface, offering dimensional stability for various decorative, shielding, and labeling applications. Designed for versatile use, it features a durable construction suitable for indoor and outdoor environments.
  • Features an aggressive, long-lasting acrylic adhesive
  • Offers excellent chemical and thermal stability
  • Includes a brilliant, vibrant, mirror-like finish
  • Compatible with a wide range of surfaces including metal, polycarbonate, and powder-coated paints
  • Withstands temperatures from -60°F to +300°F
  • Provides a tensile strength of 24 lbs./in.