Learn More
Grainger Metalized Film Tape 72 yd L 4 W
Catalog No. 502752830
Encompass_Preferred
The TAPECASE Single-Sided Splicing Tape is a 1.88 inch x 60 yard roll of splicing tape ideal for film, paper, foil, and other substrates. It features a rubber adhesive that provides quick stick and holds well. The tape has a tensile strength of 25 lbs/in width and adheres to most surfaces including paper, plastic, glass, and more. It can withstand temperatures up to 200°F. The tape tears easily by hand without curling and conforms well to uneven surfaces. It is made in the USA by TapeCase, a company focused on specialty tapes for over 70 years. This splicing tape delivers reliable holding power in a variety of industrial and commercial applications.
Catalog No. 50-275-2830 Supplier Grainger Supplier No. 15D537
The TAPECASE Single-Sided Splicing Tape is a 1.88 inch x 60 yard roll of splicing tape ideal for film, paper, foil, and other substrates. It features a rubber adhesive that provides quick stick and holds well. The tape has a tensile strength of 25 lbs/in width and adheres to most surfaces including paper, plastic, glass, and more. It can withstand temperatures up to 200°F. The tape tears easily by hand without curling and conforms well to uneven surfaces. It is made in the USA by TapeCase, a company focused on specialty tapes for over 70 years. This splicing tape delivers reliable holding power in a variety of industrial and commercial applications.