Learn More
Uline Insulated Mailers - 7 x 10 | Metallic | 25
Supplier:  Uline S22599
Encompass_Preferred
Keep chocolates medicines and perishables cold during transport, Thick padded mailers cushion and insulate up to 36 hours, Dual closure with leakproof zipper inside and exterior self-seal strip, 12 very thick foam with metalized poly exterior, FDA compliant, Use with Cold Packs
Catalog No. NC2177316