Learn More
Enzo Life Sciences Metallothionein (human) clonal antibody (N11-G) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX810014R100
Encompass_Preferred
Clone: N11-G. Isotype: IgG. Immunogen: Synthetic peptide corresponding to the N-terminal sequence of human metallothionein (covering isoforms 1A, B, E, F, G, H, I, L, M, Q, S, Y, 2A). Species reactivity: Human. Applications: ELISA, WB. Purity: Major clone of IgGs obtained from immunoaffinity purified immunoglobulins corresponding to immunogenic peptide. Formulation: Liquid. In 20mM TRIS/HCl, pH 8.0, containing 10mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-0074