Learn More
Enzo Life Sciences Metallothionein monoclonal antibody (UC1MT) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA550F
Encompass_Preferred
Clone: UC1MT. Host: Mouse. Isotype: IgG1. Immunogen: Cross-linked rabbit liver Metallothionein I and II. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Rabbit, Horse, Fish. Applications: ELISA, IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-667-479