Learn More
Electron Microscopy Sciences Thin Section Holder 3.2 mm (1⁄8") x 8mm
SDP
Catalog No. 501901527
Encompass
This mount holder which is made from machined aluminum with stainless steel Allen screws is specifically for 1" or 25 mm metallurgical mounts.
Catalog No. 50-190-1527 Supplier Electron Microscopy Sciences Supplier No. 7594105
This mount holder which is made from machined aluminum with stainless steel Allen screws is specifically for 1" or 25 mm metallurgical mounts.