Learn More
Electron Microscopy Sciences Metallurgical Mount Holder Hitachi M4
SDP
Catalog No. 501901528
Encompass
This mount holder which is made from machined aluminum with stainless steel Allen screws is specifically for 1" or 25 mm metallurgical mounts.
Catalog No. 50-190-1528 Supplier Electron Microscopy Sciences Supplier No. 7594105H
This mount holder which is made from machined aluminum with stainless steel Allen screws is specifically for 1" or 25 mm metallurgical mounts.