Learn More
Electron Microscopy Sciences Metallurgical Mount Holder 1" and 30 mm, Hitachi M4
SDP
Catalog No. 501901530
Encompass
This mount holder which is made from machined aluminum with stainless steel Allen screws. Desgned specifically for use with 1 1/4" and 30 mm metallurgical mounts.
Type: Hitachi M4
Catalog No. 50-190-1530 Supplier Electron Microscopy Sciences Supplier No. 7594106H
This mount holder which is made from machined aluminum with stainless steel Allen screws. Desgned specifically for use with 1 1/4" and 30 mm metallurgical mounts.
Type: Hitachi M4