Learn More
Electron Microscopy Sciences Metallurgical Mount Holder 1" and 30 mm, 3.2 mm x 8mm Pin
SDP
Catalog No. 501901529
Encompass
This mount holder which is made from machined aluminum with stainless steel Allen screws. Desgned specifically for use with 1 1/4" and 30 mm metallurgical mounts.
Type: 3.2 mm x 8 mm Pin
Catalog No. 50-190-1529 Supplier Electron Microscopy Sciences Supplier No. 7594106
This mount holder which is made from machined aluminum with stainless steel Allen screws. Desgned specifically for use with 1 1/4" and 30 mm metallurgical mounts.
Type: 3.2 mm x 8 mm Pin