Learn More
Electron Microscopy Sciences Metallurgical Mount Holder 2" and 50 mm, Hitachi M4
SDP
Catalog No. 501901533
Encompass
This mount holder which is made from machined aluminum with stainless steel Allen screws. Designed specifically for 2" and 50 mm metallurgical mounts.
Type: Hitachi M4
Catalog No. 50-190-1533 Supplier Electron Microscopy Sciences Supplier No. 7594109
This mount holder which is made from machined aluminum with stainless steel Allen screws. Designed specifically for 2" and 50 mm metallurgical mounts.
Type: Hitachi M4