Learn More
Electron Microscopy Sciences Metallurgical Mount Holder 1 1/2" and 40 mm, Hitachi M4
SDP
Catalog No. 501901532
Encompass
This set screw vice which is made from machined aluminum with stainless steel allen set screws has an open slot that is 10 mm (.394") wide x 5 mm (.197") deep. It measures 25 mm (1”) dia. x 17.5 mm (0.69”) high.
Type: Hitachi M4
Catalog No. 50-190-1532 Supplier Electron Microscopy Sciences Supplier No. 7594108H
This set screw vice which is made from machined aluminum with stainless steel allen set screws has an open slot that is 10 mm (.394") wide x 5 mm (.197") deep. It measures 25 mm (1”) dia. x 17.5 mm (0.69”) high.
Type: Hitachi M4