Learn More
Electron Microscopy Sciences Metallurgical Mount Holder 2" and 50 mm, 3.2 mm x 8mm Pin
SDP
Catalog No. 501901534
Encompass
This mount holder which is made from machined aluminum with stainless steel Allen screws. Designed specifically for 2" and 50 mm metallurgical mounts.
Type: 3.2 mm x 8 mm Pin
Catalog No. 50-190-1534 Supplier Electron Microscopy Sciences Supplier No. 7594109H
This mount holder which is made from machined aluminum with stainless steel Allen screws. Designed specifically for 2" and 50 mm metallurgical mounts.
Type: 3.2 mm x 8 mm Pin