Learn More
Electron Microscopy Sciences Metallurgical Graticle G41, 19mm, ASTM Austenite 1:1
SDP
Catalog No. 5027142
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM grain size austenite, glass sandwich.
Size: 19mm
Method: ASTM Austenite 1:1
Surface: Glass sandwich
Catalog No. 50-271-42 Supplier Electron Microscopy Sciences Supplier No. 6806519
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM grain size austenite, glass sandwich.
Size: 19mm
Method: ASTM Austenite 1:1
Surface: Glass sandwich