Learn More
Electron Microscopy Sciences Metallurgical Graticle G42, 19mm, ASTM E112
SDP
Catalog No. 5027151
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. Grain sizing (E112) Glass Sandwich.
Size: 19mm
Method: ASTM E112
Surface: Glass sandwich
Catalog No. 50-271-51 Supplier Electron Microscopy Sciences Supplier No. 6806619
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. Grain sizing (E112) Glass Sandwich.
Size: 19mm
Method: ASTM E112
Surface: Glass sandwich