Learn More
Electron Microscopy Sciences Metallurgical Graticle G42, 21mm, ASTM E112
SDP
Catalog No. 5027147
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. Grain sizing (E112), Glass Sandwich.
Size: 21mm
Method: ASTM E112
Surface: Glass sandwich
Catalog No. 50-271-47 Supplier Electron Microscopy Sciences Supplier No. 6806621
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. Grain sizing (E112), Glass Sandwich.
Size: 21mm
Method: ASTM E112
Surface: Glass sandwich