Learn More
Electron Microscopy Sciences Metallurgical Graticle G43, 19mm, ASTM Carbide
SDP
Catalog No. 5027139
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. Grain sizing (E112) glass sandwich.
Size: 19mm
Method: ASTM Carbide
Surface: Glass sandwich
Catalog No. 50-271-39 Supplier Electron Microscopy Sciences Supplier No. 6806719
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. Grain sizing (E112) glass sandwich.
Size: 19mm
Method: ASTM Carbide
Surface: Glass sandwich