Learn More
Electron Microscopy Sciences Metallurgical Graticle G43, 21mm, ASTM Carbide
SDP
Catalog No. 5027138
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. Grain sizing (E112) glass sandwich.
Size: 21mm
Method: ASTM Carbide
Surface: Glass sandwich
Catalog No. 50-271-38 Supplier Electron Microscopy Sciences Supplier No. 6806721
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. Grain sizing (E112) glass sandwich.
Size: 21mm
Method: ASTM Carbide
Surface: Glass sandwich