Learn More
Electron Microscopy Sciences Metallurgical Graticle G44, 21mm, ASTM E45
SDP
Catalog No. 5027155
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM E45, Root 2 sides, 7.1mm square, 10 mm scale.
Size: 21mm
Method: ASTM E45
Catalog No. 50-271-55 Supplier Electron Microscopy Sciences Supplier No. 6806821
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM E45, Root 2 sides, 7.1mm square, 10 mm scale.
Size: 21mm
Method: ASTM E45