Learn More
Electron Microscopy Sciences Metallurgical Graticle G45, 19mm, ASTM E19-46
SDP
Catalog No. 5027146
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM grain sizing E19-46, glass sandwich.
Size: 19mm
Method: ASTM E19-46
Surface: Glass sandwich
Catalog No. 50-271-46 Supplier Electron Microscopy Sciences Supplier No. 6806919
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM grain sizing E19-46, glass sandwich.
Size: 19mm
Method: ASTM E19-46
Surface: Glass sandwich