Learn More
Electron Microscopy Sciences Metallurgical Graticle G45, 21mm, ASTM E19-46
SDP
Catalog No. 5027153
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM grain sizing E19-46, glass sandwich.
Size: 21mm
Method: ASTM E19-46
Surface: Glass sandwich
Catalog No. 50-271-53 Supplier Electron Microscopy Sciences Supplier No. 6806921
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM grain sizing E19-46, glass sandwich.
Size: 21mm
Method: ASTM E19-46
Surface: Glass sandwich