Learn More
Electron Microscopy Sciences Metallurgical Graticle G46, 19mm, ASTM E19-46 Root 2
SDP
Catalog No. 5027140
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections.
Size: 19mm
Method: ASTM E19-46 Root 2
Catalog No. 50-271-40 Supplier Electron Microscopy Sciences Supplier No. 6807019
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections.
Size: 19mm
Method: ASTM E19-46 Root 2