Learn More
Electron Microscopy Sciences Metallurgical Graticle G46, 21mm, ASTM E19-46 Root 2
SDP
Catalog No. 5027154
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM grain sizing E19-46, root 2, glass sandwich.
Size: 21mm
Method: ASTM E19-46 Root 2
Surface: Glass sandwich
Catalog No. 50-271-54 Supplier Electron Microscopy Sciences Supplier No. 6807021
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM grain sizing E19-46, root 2, glass sandwich.
Size: 21mm
Method: ASTM E19-46 Root 2
Surface: Glass sandwich