Learn More
Electron Microscopy Sciences Metallurgical Graticle G54, 16mm, ASTM 24
SDP
Catalog No. 5027145
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM 24 point circular grid, glass sandwich.
Size: 16mm
Method: ASTM 24
Surface: Glass sandwich
Catalog No. 50-271-45 Supplier Electron Microscopy Sciences Supplier No. 6808416
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM 24 point circular grid, glass sandwich.
Size: 16mm
Method: ASTM 24
Surface: Glass sandwich