Learn More
Electron Microscopy Sciences Metallurgical Graticle G54, 19mm, ASTM 24
SDP
Catalog No. 5027148
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM 24 point circular grid, glass sandwich.
Size: 19mm
Method: ASTM 24
Surface: Glass sandwich
Catalog No. 50-271-48 Supplier Electron Microscopy Sciences Supplier No. 6808419
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM 24 point circular grid, glass sandwich.
Size: 19mm
Method: ASTM 24
Surface: Glass sandwich