Learn More
Electron Microscopy Sciences Metallurgical Graticle G55, 16mm, ASTM 25
SDP
Catalog No. 5027143
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM 25 point square grid, glass sandwich.
Size: 16mm
Method: ASTM 25
Surface: Glass sandwich
Catalog No. 50-271-43 Supplier Electron Microscopy Sciences Supplier No. 6808516
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM 25 point square grid, glass sandwich.
Size: 16mm
Method: ASTM 25
Surface: Glass sandwich