Learn More
Electron Microscopy Sciences Metallurgical Graticle G55, 19mm, ASTM 25
SDP
Catalog No. 5027137
Encompass_Preferred
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM 25 point square grid, glass sandwich.
Size: 19mm
Method: ASTM 25
Surface: Glass sandwich
Catalog No. 50-271-37 Supplier Electron Microscopy Sciences Supplier No. 6808519
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections. ASTM 25 point square grid, glass sandwich.
Size: 19mm
Method: ASTM 25
Surface: Glass sandwich