Learn More
Cayman Chemical MetalothIonIncod Polyclo 500uL
SDP
Catalog No. 502965598
Encompass_Preferred
A selective inhibitor of TGF-B type 1 receptor (TGFBR1 or ALK5 IC50 18 nM) inhibits the expression of collagen type I in cells (IC50 93 nM) and in mice when given orally at 10 mg/kg once a day reduces typical features of fibrosis
Catalog No. 50-296-5598 Supplier Cayman Chemical Supplier No. 172130500 UL
Add to Cart
Add to Cart
A selective inhibitor of TGF-B type 1 receptor (TGFBR1 or ALK5 IC50 18 nM) inhibits the expression of collagen type I in cells (IC50 93 nM) and in mice when given orally at 10 mg/kg once a day reduces typical features of fibrosis