Learn More
Hach Company Metals Quality Control Standard for Drinking Water, High Range, 500 mL
Supplier:  Hach Company 2833649
Encompass
Ready to use - no dilutions required. Mixed standard includes: Copper: 2.5 mg/L Cu, Iron: 1.5 mg/L Fe, Manganese: 5 mg/L Mn. 500 mL Bottle
Catalog No. 50-199-6741