Learn More
Inorganic Ventures ICP PRECIOUS METALS STD 125mL
SDP
Catalog No. 50723176
Encompass_Preferred
Matrix: Hydrochloric Acid (HCl), Analyte: Au, Pd, Rh, Ir, Pt, and Ru 100 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-176 Supplier Inorganic Ventures Supplier No. CCS2125ML
Matrix: Hydrochloric Acid (HCl), Analyte: Au, Pd, Rh, Ir, Pt, and Ru 100 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search