Learn More
MSC Rochester Midland Corporation Coolant, InTec Combo #1002, Heavy Duty, Semi-Synthetic, 55 gal
Catalog No. 502835776
Encompass
InTec Combo #1002 is a versatile medium to heavy duty semi-synthetic micro emulsion coolant developed for the metalworking industry. It is formulated to offer excellent extreme pressure characteristics, superior wetting capability, and enhanced rust protection. This coolant also resists rancidity and is designed to be mild to the operator's skin.
  • High performance, general purpose coolant
  • Exceptional cooling and EP properties
  • Excellent rust protection
  • Low foaming characteristics
  • Prolonged tool and coolant life
  • Superior resistance to rancidity
  • High operator acceptance
  • Economical and easy to use
Catalog No. 50-283-5776 Supplier MSC Supplier No. 54708615
InTec Combo #1002 is a versatile medium to heavy duty semi-synthetic micro emulsion coolant developed for the metalworking industry. It is formulated to offer excellent extreme pressure characteristics, superior wetting capability, and enhanced rust protection. This coolant also resists rancidity and is designed to be mild to the operator's skin.
  • High performance, general purpose coolant
  • Exceptional cooling and EP properties
  • Excellent rust protection
  • Low foaming characteristics
  • Prolonged tool and coolant life
  • Superior resistance to rancidity
  • High operator acceptance
  • Economical and easy to use