Learn More
Electron Microscopy Sciences Metamil, 0.25% Stock Solution 1 LT
SDP
Catalog No. 5027429
Encompass_Preferred
Mucicarmine solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is for use with hematoxylin and metanil yellow for the determination of carbohydrates and mucosubstances.
Catalog No. 50-274-29 Supplier Electron Microscopy Sciences Supplier No. 2608106
Mucicarmine solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is for use with hematoxylin and metanil yellow for the determination of carbohydrates and mucosubstances.