Learn More
AdipoGen Metamitron
SDP
Supplier:  AdipoGen CDXM0409M100
Encompass_Preferred
Chemical. CAS 41394-05-2. Formula C10H10N4O. MW 202.2. Metamitron, a chemical synthetic triazinone herbicide, is a photosystem II PSII electron transport inhibitor. It is applied as a foliar spray, and is absorbed effectively through the leaf surface and the roots. It is effective against grass and broad-leaved weeds in beet crops. Compound can be used as analytical reference material.
Catalog No. 50-256-0466