Learn More
Selleck Chemical LLC Metamizole sodium hydrate S4435-25mg
SDP
Catalog No. 503109494
Encompass_Preferred
Metamizole (Dipyrone) sodium hydrate is a potent inhitior of cyclooxygenase (COX) with strong analgesic as well as antipyretic and spasmolytic properties
Catalog No. 50-310-9494 Supplier Selleck Chemical LLC Supplier No. S443525MG
Metamizole (Dipyrone) sodium hydrate is a potent inhitior of cyclooxygenase (COX) with strong analgesic as well as antipyretic and spasmolytic properties