Learn More
Cayman Chemical Metanephrine 10mg
SDP
Catalog No. 502972897
Encompass_Preferred
An inactive metabolite of epinephrine; urinary and plasma levels are increased in patients with pheochromocytoma
Catalog No. 50-297-2897 Supplier Cayman Chemical Supplier No. 1540810 MG
An inactive metabolite of epinephrine; urinary and plasma levels are increased in patients with pheochromocytoma