Learn More
Cayman Chemical Metanephrine, Purity: ≥95%, CAS: 5001-33-2, 50 mg
SDP
Catalog No. 502971270
Encompass_Preferred
An inactive metabolite of epinephrine; urinary and plasma levels are increased in patients with pheochromocytoma
Catalog No. 50-297-1270 Supplier Cayman Chemical Supplier No. 1540850 MG
An inactive metabolite of epinephrine; urinary and plasma levels are increased in patients with pheochromocytoma