Learn More
Electron Microscopy Sciences Metanil Yellow 0.25% 500 ML
SDP
Catalog No. 5030071
Encompass_Preferred
Used in Mayer Mucicarmine Method for mucosubstances stains.
Catalog No. 50-300-71 Supplier Electron Microscopy Sciences Supplier No. 2632002
Used in Mayer Mucicarmine Method for mucosubstances stains.