Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metanil Yellow, MP Biomedicals

Catalog No. ICN211942 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211942 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211942 Supplier MP Biomedicals, Inc Supplier No. 0521194280
Add to Cart
Add to Cart

Chemical Identifiers

CAS 587-98-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.378
InChI Key NYGZLYXAPMMJTE-UHFFFAOYSA-M
Synonym metanil yellow, acid yellow 36, acid metanil yellow, monoazo, amacid yellow m, fenazo yellow m, acid golden g, kiton yellow ms, metanil yellow c, metanil yellow e
PubChem CID 3935589
ChEBI CHEBI:87235
IUPAC Name sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 100 g
Formula Weight 375.38
Chemical Name or Material Metanil Yellow
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.