Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Metanil Yellow (Tech.), 85%

Catalog No. AC413721000
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC413721000 100 g
AC413722500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413721000 Supplier Thermo Scientific Chemicals Supplier No. 413721000
Add to Cart

CAS: 587-98-4 | C18H14N3NaO3S | 375.378 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-98-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.378
InChI Key NYGZLYXAPMMJTE-UHFFFAOYSA-M
Synonym Acid Yellow 36, 3-[[4-(Phenylamino)phenyl]azo]benzenesulfonic acid monosodium salt, C.I. 13065, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
PubChem CID 3935589
ChEBI CHEBI:87235
IUPAC Name sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]

Specifications

Dye Content 80% min.
Beilstein 16, 330
Merck Index 15, 5999
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol, moderately soluble in benzene and ether, slightly soluble in acetone
Infrared Spectrum Authentic
Formula Weight 375.38
Grade Pure
Packaging Glass bottle
Color Orange
Quantity 100 g
Physical Form Powder
Chemical Name or Material Metanil Yellow, tech., 85%
Show More Show Less
EINECSNumber 209-608-2
RTECSNumber DB7329500
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.