Learn More
Aviva Systems Biology Corp METAP1 Antibody - N-terminal region (ARP55179_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55179_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against METAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human METAP1. Alternate names: DKFZp781C0419, KIAA0094, MAP1A, MetAP1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61433