Learn More
Medchemexpress LLC Methionine aminopeptidase 2 protein, human (Sf9, His) | >95.0% | 500UG
SDP
Catalog No. 5000175061
Encompass_Preferred
Recombinant human METAP2 (methionine aminopeptidase 2) expressed in Sf9 insect cells with an N-terminal 6xHis tag. Supplied as a 0.22 μm-filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0, the protein is activity-validated and shipped frozen for long-term storage.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~58-70 kDa due to glycosylation.
  • Enzymatically active, specific activity >15 pmol/min/μg.
  • Ready-to-use liquid formulation, 0.22 μm filtered.
  • Suitable for biochemical assays and enzyme characterization.
  • Shipped on dry ice; store at -80°C to preserve activity.
  • N-terminal His tag facilitates purification and detection.
Catalog No. 50-001-75061 Supplier Medchemexpress LLC Supplier No. HYP70364500UG
Recombinant human METAP2 (methionine aminopeptidase 2) expressed in Sf9 insect cells with an N-terminal 6xHis tag. Supplied as a 0.22 μm-filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0, the protein is activity-validated and shipped frozen for long-term storage.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~58-70 kDa due to glycosylation.
  • Enzymatically active, specific activity >15 pmol/min/μg.
  • Ready-to-use liquid formulation, 0.22 μm filtered.
  • Suitable for biochemical assays and enzyme characterization.
  • Shipped on dry ice; store at -80°C to preserve activity.
  • N-terminal His tag facilitates purification and detection.