Learn More
Medchemexpress LLC Metaphosphoric acid | 37267-86-0 | MFCD00011351 | 10g
SDP
Catalog No. 5000266259
Encompass_Preferred
Metaphosphoric acid 33 5-36 5% (Glacial phosphoric acid 33 5-36 5%) is a precipitant for rapid enzymatic measurement of blood lactate and pyruvate[1]
Catalog No. 50-002-66259 Supplier Medchemexpress LLC Supplier No. HYY1939A10G
Metaphosphoric acid 33 5-36 5% (Glacial phosphoric acid 33 5-36 5%) is a precipitant for rapid enzymatic measurement of blood lactate and pyruvate[1]