Learn More
Cayman Chemical Metapramine, Purity: ≥98%, CAS: 21730-16-5, 10 mg
SDP
Catalog No. 502978387
Encompass_Preferred
An antidepressant; decreases action potential amplitude and maximal signal velocity and increases conduction time in isolated dog Purkinje fibers at 10 µM; reduces ethanol consumption in ethanol-preferring rats at 10 mg/kg; inhibits reperfusion-induced increases in ventricular arrhythmias in a rat model of myocardial ischemia-reperfusion injury induced by ligation of the LAD artery at 175 µM; increases the latency to tail withdrawal in the tail-flick test and inhibits phenylbenzoquinone-induced writhing in mice at 10 mg/kg
Catalog No. 50-297-8387 Supplier Cayman Chemical Supplier No. 4203310 MG
An antidepressant; decreases action potential amplitude and maximal signal velocity and increases conduction time in isolated dog Purkinje fibers at 10 µM; reduces ethanol consumption in ethanol-preferring rats at 10 mg/kg; inhibits reperfusion-induced increases in ventricular arrhythmias in a rat model of myocardial ischemia-reperfusion injury induced by ligation of the LAD artery at 175 µM; increases the latency to tail withdrawal in the tail-flick test and inhibits phenylbenzoquinone-induced writhing in mice at 10 mg/kg