Learn More
Cayman Chemical Metapristone, Purity: ≥98%, CAS: 104004-96-8, 10 mg
SDP
Catalog No. 502911737
Encompass_Preferred
An active metabolite of mifepristone; an antagonist of human glucocorticoid and progesterone receptors with relative affinities of 61 and 21%, respectively, compared to mifepristone; decreases the proliferation of A375 and B16/F10 cells (IC50s = 57.4 and 75.8 µM, respectively); induces cell-cycle arrest at the G1 phase in B16/F10 cells at 80 µM; decreases the number of lung metastases in a B16/F10 murine model of lung metastasis at 2.5, 10, and 50 mg/kg per day
Catalog No. 50-291-1737 Supplier Cayman Chemical Supplier No. 3638010 MG
An active metabolite of mifepristone; an antagonist of human glucocorticoid and progesterone receptors with relative affinities of 61 and 21%, respectively, compared to mifepristone; decreases the proliferation of A375 and B16/F10 cells (IC50s = 57.4 and 75.8 µM, respectively); induces cell-cycle arrest at the G1 phase in B16/F10 cells at 80 µM; decreases the number of lung metastases in a B16/F10 murine model of lung metastasis at 2.5, 10, and 50 mg/kg per day