Learn More
Medchemexpress LLC Metaproterenol (hemisulfate) | 5874-97-5 | 99.4% | 100 MG
SDP
Catalog No. 5000411873
Encompass_Preferred
Metaproterenol hemisulfate (Orciprenaline hemisulfate) is a direct-acting sympathomimetic and a β2-adrenergic receptor (β2AR) agonist with an IC50 of 68 nM. It also exhibits anti-inflammatory activity.
  • Functions as a β2-adrenergic receptor (β2AR) agonist with an IC50 of 68 nM.
  • Possesses anti-inflammatory activity.
  • Enhances β-arrestin2 and its interaction with IκBα in high glucose-induced THP-1 cells and bone marrow macrophages.
  • Downregulates NF-κB in high glucose-induced THP-1 cells and bone marrow macrophages.
  • Attenuates monocyte activation and pro-inflammatory and pro-fibrotic responses in the kidneys and heart of Zucker diabetic fatty rats, potentially offering protective effects against diabetic renal and cardiovascular complications.
Catalog No. 50-004-11873 Supplier Medchemexpress LLC Supplier No. HYB1276100MG
Metaproterenol hemisulfate (Orciprenaline hemisulfate) is a direct-acting sympathomimetic and a β2-adrenergic receptor (β2AR) agonist with an IC50 of 68 nM. It also exhibits anti-inflammatory activity.
  • Functions as a β2-adrenergic receptor (β2AR) agonist with an IC50 of 68 nM.
  • Possesses anti-inflammatory activity.
  • Enhances β-arrestin2 and its interaction with IκBα in high glucose-induced THP-1 cells and bone marrow macrophages.
  • Downregulates NF-κB in high glucose-induced THP-1 cells and bone marrow macrophages.
  • Attenuates monocyte activation and pro-inflammatory and pro-fibrotic responses in the kidneys and heart of Zucker diabetic fatty rats, potentially offering protective effects against diabetic renal and cardiovascular complications.