Learn More
Medchemexpress LLC Metaproterenol hemisulfate | 5874-97-5 | MFCD00058303 | 99.4% | 1 ML
SDP
Catalog No. 5000412822
Encompass_Preferred
Metaproterenol hemisulfate from MedChemExpress is a direct-acting sympathomimetic and a β2-adrenergic receptor (β2AR) agonist. It also exhibits anti-inflammatory activity and is for research use only.
  • Direct-acting sympathomimetic
  • β2-adrenergic receptor (β2AR) agonist
  • Exhibits anti-inflammatory activity
  • Enhances β-arrestin2 and its interaction with IκBα
  • Downregulates NF-κB
  • Attenuates monocyte activation and pro-inflammatory and pro-fibrotic responses in the kidneys and heart
  • Potential protective effects against diabetic renal and cardiovascular complications
Catalog No. 50-004-12822 Supplier Medchemexpress LLC Supplier No. HYB127610MM/1ML
Metaproterenol hemisulfate from MedChemExpress is a direct-acting sympathomimetic and a β2-adrenergic receptor (β2AR) agonist. It also exhibits anti-inflammatory activity and is for research use only.
  • Direct-acting sympathomimetic
  • β2-adrenergic receptor (β2AR) agonist
  • Exhibits anti-inflammatory activity
  • Enhances β-arrestin2 and its interaction with IκBα
  • Downregulates NF-κB
  • Attenuates monocyte activation and pro-inflammatory and pro-fibrotic responses in the kidneys and heart
  • Potential protective effects against diabetic renal and cardiovascular complications